3-(3-Methylphenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech

Lieferant: Apollo Scientific

OR33280-1G
APOSOR33280-1GEA 1570 EUR
APOSOR33280-1G
3-(3-Methylphenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
3-(3-Methylphenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Formel: C₁₈H₁₇NO₃
Molecular Weight: 295,332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571408
CAS-Nummer: 478260-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR