(2Z)-2-[(Pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Lieferant: Apollo Scientific

OR33283-1G
APOSOR33283-1GEA 1570 EUR
APOSOR33283-1G
(2Z)-2-[(Pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(Pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₃H₁₄N₂O
Molecular Weight: 214,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845415
CAS-Nummer: 410087-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR