(2Z)-2-[(Pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech

Lieferant: Apollo Scientific

OR33284-1G
APOSOR33284-1GEA 1570 EUR
APOSOR33284-1G
(2Z)-2-[(Pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech
(2Z)-2-[(Pyridin-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol
Formel: C₁₃H₁₆N₂O
Molecular Weight: 216,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845414
CAS-Nummer: 866136-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR