3-Cyclohexyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech

Lieferant: Apollo Scientific

OR33285-1G
APOSOR33285-1GEA 1570 EUR
APOSOR33285-1G
3-Cyclohexyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
3-Cyclohexyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Formel: C₁₇H₂₁NO₃
Molecular Weight: 287,354 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305813
CAS-Nummer: 866137-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR