7-Chloro-6-(2-chloroethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine, tech

Lieferant: Apollo Scientific

OR33287-1G
APOSOR33287-1GEA 1570 EUR
APOSOR33287-1G
7-Chloro-6-(2-chloroethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine, tech
7-Chloro-6-(2-chloroethyl)-2,5-dimethylpyrazolo[1,5-a]pyrimidine
Formel: C₁₀H₁₁Cl₂N₃
Molecular Weight: 244,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305817
CAS-Nummer: 129909-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR