N-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-4-carboxamide, tech

Lieferant: Apollo Scientific

OR33292-1G
APOSOR33292-1GEA 1570 EUR
APOSOR33292-1G
N-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-4-carboxamide, tech
N-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridine-4-carboxamide
Formel: C₁₂H₁₃N₃O
Molecular Weight: 215,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01717893
CAS-Nummer: 5588-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR