4-(Chloromethyl)-7-hydroxy-8-methyl-2H-chromen-2-one 95%

Lieferant: Apollo Scientific

OR33293-1G
APOSOR33293-1GEA 427 EUR
APOSOR33293-1G
4-(Chloromethyl)-7-hydroxy-8-methyl-2H-chromen-2-one 95%
4-(Chloromethyl)-7-hydroxy-8-methyl-2H-chromen-2-one
Formel: C₁₁H₉ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03937668
CAS-Nummer: 163684-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR