6-(4-Benzylpiperazino)nicotinic acid 95%

Lieferant: Apollo Scientific

Synonyme: 6-(4-Benzylpiperazin-1-yl)pyridine-3-carboxylic acid

OR33294-1G OR33294-5G
APOSOR33294-1GEA 306 EUR
APOSOR33294-1G APOSOR33294-5G
6-(4-Benzylpiperazino)nicotinic acid 95%
6-(4-Benzylpiperazino)nicotinic acid
Formel: C₁₇H₁₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03791243
CAS-Nummer: 886360-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR