N-(Prop-2-en-1-yl)pyrimidin-2-amine 95%
Lieferant: Apollo Scientific
OR33297-1G
APOSOR33297-1GEA
1120
EUR
APOSOR33297-1G
N-(Prop-2-en-1-yl)pyrimidin-2-amine 95%
N-(Prop-2-en-1-yl)pyrimidin-2-amine
Formel:
C₇H₉N₃ Molecular Weight: 135,166 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04126151 CAS-Nummer: 5176-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...