(2Z)-2-[(2,5-Dimethoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech

Lieferant: Apollo Scientific

OR33299-1G
APOSOR33299-1GEA 1570 EUR
APOSOR33299-1G
(2Z)-2-[(2,5-Dimethoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol, tech
(2Z)-2-[(2,5-Dimethoxyphenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-ol
Formel: C₁₆H₂₁NO₃
Molecular Weight: 275,343 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05256362
CAS-Nummer: 866138-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR