1-{6-[(2-Hydroxyethyl)(methyl)amino]naphthalen-2-yl}ethan-1-one 97%

Lieferant: Apollo Scientific

OR33301-1G
APOSOR33301-1GEA 1570 EUR
APOSOR33301-1G
1-{6-[(2-Hydroxyethyl)(methyl)amino]naphthalen-2-yl}ethan-1-one 97%
1-{6-[(2-Hydroxyethyl)(methyl)amino]naphthalen-2-yl}ethan-1-one
Formel: C₁₅H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12756735
CAS-Nummer: 259739-00-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR