(1R)-2-(3-Bromophenyl)cyclopropane-1-carboxylic acid 95%

Lieferant: Apollo Scientific

OR33305-1G
APOSOR33305-1GEA 1410 EUR
APOSOR33305-1G
(1R)-2-(3-Bromophenyl)cyclopropane-1-carboxylic acid 95%
(1R)-2-(3-Bromophenyl)cyclopropane-1-carboxylic acid
Formel: C₁₀H₉BrO₂
Molecular Weight: 241,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28805784
CAS-Nummer: 1314323-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR