2-Azaspiro[4.4]nonan-3-one 95%

Lieferant: Apollo Scientific

OR33313-1G OR33313-5G
APOSOR33313-1GEA 78.3 EUR
APOSOR33313-1G APOSOR33313-5G
2-Azaspiro[4.4]nonan-3-one 95%
2-Azaspiro[4.4]nonan-3-one
Formel: C₈H₁₃NO
Molecular Weight: 139.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864498
CAS-Nummer: 75751-72-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR