N-tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxamide, tech

Lieferant: Apollo Scientific

OR33318-1G
APOSOR33318-1GEA 1570 EUR
APOSOR33318-1G
N-tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxamide, tech
N-tert-Butyl-4-(6-chloropyridin-2-yl)piperazine-1-carboxamide
Formel: C₁₄H₂₁ClN₄O
Molecular Weight: 296,796 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139041
CAS-Nummer: 339107-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR