(4E)-4-[(Dimethylamino)methylidene]-2,2-diphenyloxolan-3-one, tech

Lieferant: Apollo Scientific

OR33319-1G
APOSOR33319-1GEA 1570 EUR
APOSOR33319-1G
(4E)-4-[(Dimethylamino)methylidene]-2,2-diphenyloxolan-3-one, tech
(4E)-4-[(Dimethylamino)methylidene]-2,2-diphenyloxolan-3-one
Formel: C₁₉H₁₉NO₂
Molecular Weight: 293.36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127630
CAS-Nummer: 339107-43-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR