(2E)-3-(Dimethylamino)-1-[4-(4-methylpiperazin-1-yl)phenyl]prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33323-1G
APOSOR33323-1GEA 1570 EUR
APOSOR33323-1G
(2E)-3-(Dimethylamino)-1-[4-(4-methylpiperazin-1-yl)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[4-(4-methylpiperazin-1-yl)phenyl]prop-2-en-1-one
Formel: C₁₆H₂₃N₃O
Molecular Weight: 273,373 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138783
CAS-Nummer: 338975-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR