Methyl-1-chloro-4-isoquinolinecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-chloroisoquinoline-4-carboxylate

OR33324-1G
APOSOR33324-1GEA 857 EUR
APOSOR33324-1G
Methyl-1-chloro-4-isoquinolinecarboxylate 95%
Methyl-1-chloro-4-isoquinolinecarboxylate
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140353
CAS-Nummer: 37497-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR