4-[4-(4-Methylpiperazin-1-yl)phenyl]pyrimidin-2-amine, tech

Lieferant: Apollo Scientific

OR33325-1G
APOSOR33325-1GEA 1570 EUR
APOSOR33325-1G
4-[4-(4-Methylpiperazin-1-yl)phenyl]pyrimidin-2-amine, tech
4-[4-(4-Methylpiperazin-1-yl)phenyl]pyrimidin-2-amine
Formel: C₁₅H₁₉N₅
Molecular Weight: 269,345 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00138785
CAS-Nummer: 338975-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR