2-(5-Isoxazolyl)phenol 95%

Lieferant: Apollo Scientific

OR33328-5G
APOSOR33328-5GEA 0 EUR
APOSOR33328-5G
2-(5-Isoxazolyl)phenol 95%
2-(5-Isoxazolyl)phenol
Formel: C₉H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00218653
CAS-Nummer: 61348-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR