4-Acetamidophenylacetonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-(Cyanomethyl)acetanilide, N-(4-(Cyanomethyl)phenyl)acetamide

OR3333-1G OR3333-5G
APOSOR3333-1GEA 72.5 EUR
APOSOR3333-1G APOSOR3333-5G
4-Acetamidophenylacetonitrile
4-Acetamidophenylacetonitrile
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.2 g/mol
Schmelzpunkt: 82…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016398
CAS-Nummer: 25025-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR