1-(4-Acetylphenyl)-1,4-dihydropyridin-4-one, tech

Lieferant: Apollo Scientific

OR33333-1G
APOSOR33333-1GEA 1570 EUR
APOSOR33333-1G
1-(4-Acetylphenyl)-1,4-dihydropyridin-4-one, tech
1-(4-Acetylphenyl)-1,4-dihydropyridin-4-one
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139359
CAS-Nummer: 338976-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR