4-Phenylpiperazine-1-carboxamide, tech

Lieferant: Apollo Scientific

OR33334-1G
APOSOR33334-1GEA 1570 EUR
APOSOR33334-1G
4-Phenylpiperazine-1-carboxamide, tech
4-Phenylpiperazine-1-carboxamide
Formel: C₁₁H₁₅N₃O
Molecular Weight: 205.256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214814
CAS-Nummer: 77464-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR