N-[(1E)-(Dimethylamino)methylidene]-4-phenylpiperazine-1-carboxamide, tech

Lieferant: Apollo Scientific

OR33336-1G
APOSOR33336-1GEA 1570 EUR
APOSOR33336-1G
N-[(1E)-(Dimethylamino)methylidene]-4-phenylpiperazine-1-carboxamide, tech
N-[(1E)-(Dimethylamino)methylidene]-4-phenylpiperazine-1-carboxamide
Formel: C₁₄H₂₀N₄O
Molecular Weight: 260,335 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202460
CAS-Nummer: 338977-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR