N-[(1Z)-(Methoxyimino)methyl]-4-phenylpiperazine-1-carboxamide, tech

Lieferant: Apollo Scientific

OR33337-1G
APOSOR33337-1GEA 1570 EUR
APOSOR33337-1G
N-[(1Z)-(Methoxyimino)methyl]-4-phenylpiperazine-1-carboxamide, tech
N-[(1Z)-(Methoxyimino)methyl]-4-phenylpiperazine-1-carboxamide
Formel: C₁₃H₁₈N₄O₂
Molecular Weight: 262,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214815
CAS-Nummer: 338977-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR