5-Chloro-1-[(3-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid, tech

Lieferant: Apollo Scientific

OR33339-1G
APOSOR33339-1GEA 1570 EUR
APOSOR33339-1G
5-Chloro-1-[(3-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid, tech
5-Chloro-1-[(3-chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid
Formel: C₁₃H₉Cl₂NO₃
Molecular Weight: 298.121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140530
CAS-Nummer: 222414-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR