4-Chloro[1,2,4]triazolo[4,3-a]quinoxaline 95%

Lieferant: Apollo Scientific

OR33340-1G
APOSOR33340-1GEA 336 EUR
APOSOR33340-1G
4-Chloro[1,2,4]triazolo[4,3-a]quinoxaline 95%
4-Chloro[1,2,4]triazolo[4,3-a]quinoxaline
Formel: C₉H₅ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00140798
CAS-Nummer: 62603-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR