Methyl (2E)-3-(dimethylamino)-2-[(E)-(phenylmethylidene)amino]prop-2-enoate, tech

Lieferant: Apollo Scientific

OR33341-1G
APOSOR33341-1GEA 1570 EUR
APOSOR33341-1G
Methyl (2E)-3-(dimethylamino)-2-[(E)-(phenylmethylidene)amino]prop-2-enoate, tech
Methyl (2E)-3-(dimethylamino)-2-[(E)-(phenylmethylidene)amino]prop-2-enoate
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141630
CAS-Nummer: 76862-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR