(E)-N,N-Dimethyl-N'-(5-methyl-1,2-oxazol-3-yl)methanimidamide 95%
Lieferant: Apollo Scientific
OR33342-1G
APOSOR33342-1GEA
1120
EUR
APOSOR33342-1G
(E)-N,N-Dimethyl-N'-(5-methyl-1,2-oxazol-3-yl)methanimidamide 95%
(E)-N,N-Dimethyl-N'-(5-methyl-1,2-oxazol-3-yl)methanimidamide
Formel:
C₇H₁₁N₃O Molecular Weight: 153,182 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00117221 CAS-Nummer: 134540-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...