(E)-(3-{[1,1'-Biphenyl]-4-yl}-3-oxopropylidene)(methyl)oxidoazanium, tech
Lieferant: Apollo Scientific
OR33344-1G
APOSOR33344-1GEA
1570
EUR
APOSOR33344-1G
(E)-(3-{[1,1'-Biphenyl]-4-yl}-3-oxopropylidene)(methyl)oxidoazanium, tech
(E)-(3-{[1,1'-Biphenyl]-4-yl}-3-oxopropylidene)(methyl)oxidoazanium
Formel:
C₁₆H₁₅NO₂ Molecular Weight: 253,296 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173339 CAS-Nummer: 339105-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...