(2Z)-4-Chloro-3-oxo-2-[(phenylamino)methylidene]butanenitrile, tech
Lieferant: Apollo Scientific
OR33348-1G
APOSOR33348-1GEA
1570
EUR
APOSOR33348-1G
(2Z)-4-Chloro-3-oxo-2-[(phenylamino)methylidene]butanenitrile, tech
(2Z)-4-Chloro-3-oxo-2-[(phenylamino)methylidene]butanenitrile
Formel:
C₁₁H₉ClN₂O Molecular Weight: 220,655 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00663751 CAS-Nummer: 339106-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...