(2Z)-2-[(1E)-{[(4-Chlorophenyl)methoxy]imino}methyl]-3-(pyridin-4-yl)prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR33349-1G
APOSOR33349-1GEA 1570 EUR
APOSOR33349-1G
(2Z)-2-[(1E)-{[(4-Chlorophenyl)methoxy]imino}methyl]-3-(pyridin-4-yl)prop-2-enenitrile, tech
(2Z)-2-[(1E)-{[(4-Chlorophenyl)methoxy]imino}methyl]-3-(pyridin-4-yl)prop-2-enenitrile
Formel: C₁₆H₁₂ClN₃O
Molecular Weight: 297,739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663756
CAS-Nummer: 339106-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR