2-Oxo-6-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33353-1G
APOSOR33353-1GEA 1570 EUR
APOSOR33353-1G
2-Oxo-6-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
2-Oxo-6-phenyl-1-(prop-2-yn-1-yl)-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₅H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00202806
CAS-Nummer: 339109-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR