6-(4-Chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR33354-1G
APOSOR33354-1GEA 1570 EUR
APOSOR33354-1G
6-(4-Chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
6-(4-Chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₃H₉ClN₂O
Molecular Weight: 244,676 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00202815
CAS-Nummer: 339109-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR