Ethyl 4-(2-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Lieferant: Apollo Scientific
OR33360-1G
APOSOR33360-1GEA
1570
EUR
APOSOR33360-1G
Ethyl 4-(2-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Ethyl 4-(2-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate
Formel:
C₁₅H₁₆ClNO₃ Molecular Weight: 293,745 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975282 CAS-Nummer: 176383-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...