Methyl 4-(3-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

OR33361-1G
APOSOR33361-1GEA 1570 EUR
APOSOR33361-1G
Methyl 4-(3-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate, tech
Methyl 4-(3-chlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate
Formel: C₁₄H₁₄ClNO₃
Molecular Weight: 279,719 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975285
CAS-Nummer: 303140-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR