3-Chloro-N-(4-cyanophenyl)-2,2-dimethylpropanamide, tech

Lieferant: Apollo Scientific

OR33363-1G
APOSOR33363-1GEA 1570 EUR
APOSOR33363-1G
3-Chloro-N-(4-cyanophenyl)-2,2-dimethylpropanamide, tech
3-Chloro-N-(4-cyanophenyl)-2,2-dimethylpropanamide
Formel: C₁₂H₁₃ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00793439
CAS-Nummer: 478261-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR