4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 4-(3,3-Dimethyl-2-oxoazetidin-1-yl)benzonitrile

OR33364-1G
APOSOR33364-1GEA 671 EUR
APOSOR33364-1G
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile 95%
4-(3,3-Dimethyl-2-oxo-1-azetanyl)benzenecarbonitrile
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793440
CAS-Nummer: 454473-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR