3-Chloro-N-(4-acetamidophenyl)-2,2-dimethylpropanamide, tech

Lieferant: Apollo Scientific

OR33367-1G
APOSOR33367-1GEA 1570 EUR
APOSOR33367-1G
3-Chloro-N-(4-acetamidophenyl)-2,2-dimethylpropanamide, tech
3-Chloro-N-(4-acetamidophenyl)-2,2-dimethylpropanamide
Formel: C₁₃H₁₇ClN₂O₂
Molecular Weight: 268,739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793452
CAS-Nummer: 478261-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR