2-[4-(3,3-Dimethyl-2-oxo-1-azetanyl)phenyl]acetonitrile 95%

Lieferant: Apollo Scientific

OR33369-1G
APOSOR33369-1GEA 671 EUR
APOSOR33369-1G
2-[4-(3,3-Dimethyl-2-oxo-1-azetanyl)phenyl]acetonitrile 95%
2-[4-(3,3-Dimethyl-2-oxo-1-azetanyl)phenyl]acetonitrile
Formel: C₁₃H₁₄N₂O
Molecular Weight: 214,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793454
CAS-Nummer: 478262-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR