3,5-Dibromo-4,6-dimethylpyridin-2-amine, tech

Lieferant: Apollo Scientific

OR33370-1G
APOSOR33370-1GEA 398 EUR
APOSOR33370-1G
3,5-Dibromo-4,6-dimethylpyridin-2-amine, tech
3,5-Dibromo-4,6-dimethylpyridin-2-amine
Formel: C₇H₈Br₂N₂
Molecular Weight: 279,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975063
CAS-Nummer: 5407-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR