2,4-Dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
Lieferant: Apollo Scientific
OR33371-1G
APOSOR33371-1GEA
1570
EUR
APOSOR33371-1G
2,4-Dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
2,4-Dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile
Formel:
C₉H₈N₄ Molecular Weight: 172,187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975066 CAS-Nummer: 55418-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...