4-(Dimethylamino)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%

Lieferant: Apollo Scientific

OR33372-1G
APOSOR33372-1GEA 609 EUR
APOSOR33372-1G
4-(Dimethylamino)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile 95%
4-(Dimethylamino)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile
Formel: C₈H₉N₃O
Molecular Weight: 163,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01314755
CAS-Nummer: 62321-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR