2-Chloro-4-(dimethylamino)nicotinonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-4-(dimethylamino)pyridine-3-carbonitrile

OR33373-1G
APOSOR33373-1GEA 404 EUR
APOSOR33373-1G
2-Chloro-4-(dimethylamino)nicotinonitrile 95%
2-Chloro-4-(dimethylamino)nicotinonitrile
Formel: C₈H₈ClN₃
Molecular Weight: 181,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00563985
CAS-Nummer: 147992-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR