[(3-Chlorophenyl)methyl]urea 95%

Lieferant: Apollo Scientific

OR33374-1G
APOSOR33374-1GEA 0 EUR
APOSOR33374-1G
[(3-Chlorophenyl)methyl]urea 95%
[(3-Chlorophenyl)methyl]urea
Formel: C₈H₉ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00052920
CAS-Nummer: 76502-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR