Ethyl 2,5,7-trimethylimidazo[1,2-a]pyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

OR33375-1G
APOSOR33375-1GEA 1570 EUR
APOSOR33375-1G
Ethyl 2,5,7-trimethylimidazo[1,2-a]pyridine-3-carboxylate, tech
Ethyl 2,5,7-trimethylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082626
CAS-Nummer: 81438-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR