N'-[(Oxolan-3-yl)methyl](tert-butoxy)carbohydrazide, tech

Lieferant: Apollo Scientific

OR33380-1G
APOSOR33380-1GEA 1570 EUR
APOSOR33380-1G
N'-[(Oxolan-3-yl)methyl](tert-butoxy)carbohydrazide, tech
N'-[(Oxolan-3-yl)methyl](tert-butoxy)carbohydrazide
Formel: C₁₀H₂₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03305880
CAS-Nummer: 866143-73-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR