4,5-Dichloro-2-(2-oxopropyl)-2,3-dihydropyridazin-3-one, tech

Lieferant: Apollo Scientific

OR33382-1G
APOSOR33382-1GEA 1570 EUR
APOSOR33382-1G
4,5-Dichloro-2-(2-oxopropyl)-2,3-dihydropyridazin-3-one, tech
4,5-Dichloro-2-(2-oxopropyl)-2,3-dihydropyridazin-3-one
Formel: C₇H₆Cl₂N₂O₂
Molecular Weight: 221,041 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496227
CAS-Nummer: 130159-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR