3-Benzyl-1-[(furan-2-yl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR33383-1G
APOSOR33383-1GEA 1570 EUR
APOSOR33383-1G
3-Benzyl-1-[(furan-2-yl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one, tech
3-Benzyl-1-[(furan-2-yl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one
Formel: C₁₇H₁₅NO₃
Molecular Weight: 281,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871741
CAS-Nummer: 478247-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR