3-Benzyl-4-hydroxy-1-(prop-2-yn-1-yl)-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR33384-1G
APOSOR33384-1GEA 1570 EUR
APOSOR33384-1G
3-Benzyl-4-hydroxy-1-(prop-2-yn-1-yl)-1,2-dihydropyridin-2-one, tech
3-Benzyl-4-hydroxy-1-(prop-2-yn-1-yl)-1,2-dihydropyridin-2-one
Formel: C₁₅H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01871744
CAS-Nummer: 478247-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR