2-(Benzylamino)-3-chloro-1,4-dihydronaphthalene-1,4-dione, tech
Lieferant: Apollo Scientific
OR33389-1G
APOSOR33389-1GEA
1570
EUR
APOSOR33389-1G
2-(Benzylamino)-3-chloro-1,4-dihydronaphthalene-1,4-dione, tech
2-(Benzylamino)-3-chloro-1,4-dihydronaphthalene-1,4-dione
Formel:
C₁₇H₁₂ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00169805 CAS-Nummer: 4497-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...